Learn More
Epigentek Inc 5-Methylcytosine (5-mC) Monoclonal Antibody [33D3] (10 ug)
SDP
Supplier:  Epigentek Inc A1014010
Encompass_Preferred
Unconjugated Monoclonal Antibody Raised in Mouse. Immunogen: Ovalbumin-conjugated 5-methylcytosine (5-mC). Tested Applications: DB, IHC, IF, ELISA, MeDIP, IP. Reactivity: Broad Range. Alternative Names: 5-methylcytidine, anti-5-methylcytidine, anti-5-methylcytosine, anti-5mC, anti-5-mC, anti-5meC, anti-5-meC, 5mC, 5meC, 5-meC, 5'-methyl-2'-deoxycytidine, 5MedCyd.
Catalog No. 50-109-4954