Learn More
Medchemexpress LLC 5-methyldihydrofuran-2(3H)-one (gamma-valerolactone) | 108-29-2 | MFCD00005400 | 99.9% | 100.12 g/mol | C5H8O2 | 100 G
SDP
Catalog No. 5000299669
Encompass_Preferred
Gamma-valerolactone (5-methyldihydrofuran-2(3H)-one) is a colorless to light yellow liquid lactone used as a biochemical reagent and analytical reference standard for life-science and analytical applications. It has molecular formula C5H8O2, molecular weight 100.12 g/mol, and CAS 108-29-2. Intended for research use only.
  • High purity suitable for analytical applications.
  • Colorless to light yellow liquid with density ~1.05 g/cm3.
  • Versatile solvent and reagent for organic synthesis and analysis.
  • Commonly used as an analytical reference standard.
  • Stable under recommended storage conditions for extended shelf life.
Catalog No. 50-002-99669 Supplier Medchemexpress LLC Supplier No. HYW015938100G
Gamma-valerolactone (5-methyldihydrofuran-2(3H)-one) is a colorless to light yellow liquid lactone used as a biochemical reagent and analytical reference standard for life-science and analytical applications. It has molecular formula C5H8O2, molecular weight 100.12 g/mol, and CAS 108-29-2. Intended for research use only.
  • High purity suitable for analytical applications.
  • Colorless to light yellow liquid with density ~1.05 g/cm3.
  • Versatile solvent and reagent for organic synthesis and analysis.
  • Commonly used as an analytical reference standard.
  • Stable under recommended storage conditions for extended shelf life.