Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Methylfurfural

Catalog No. 1373165G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-731-65G 5 g
13-731-6100G 100 g
13-731-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-731-65G Supplier Sigma Aldrich Supplier No. 1373165G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 5-methylfurfural is formed during the photoexposition of ranitidine hydrochloride
  • It is employed as potential age marker for madeira wine
  • It is a volatile compound present in lavandula stoechas, lavandula angustifolia and lavandula angustifolia x latifolia unifloral honeys.5-methylfurfural was used to investigate the volatile compounds isolated from erica arborea and calluna vulgaris unifloral honeys.reagentplus is a registered trademark of sigma-aldrich co
  • Llc

Specifications

CAS 620-02-0
Density 1.107 g/mL (at 25°C (literature))
Boiling Point 187°C (lit.)
Molecular Formula C6H6O2
Linear Formula C6H6O2
Refractive Index n20/D 1.531 (literature)
MDL Number MFCD00003232
Quantity 5 g
Synonym 5-Methyl-2-furaldehyde
Molecular Weight (g/mol) 110.11
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.