Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylisophthalic Acid 98.0+%, TCI America™
SDP

Catalog No. M190325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M190325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M190325G Supplier TCI America Supplier No. M190325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 499-49-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00013986
InChI Key PMZBHPUNQNKBOA-UHFFFAOYSA-N
Synonym 5-methylisophthalic acid, uvitic acid, 5-methyl-1,3-benzenedicarboxylic acid, isophthalic acid, 5-methyl, 1,3-benzenedicarboxylic acid, 5-methyl, uvitinsaure, 3,5-dicarboxytoluene, 5-methylisophthalicacid, acmc-209kj2, ksc237e4t
PubChem CID 68137
IUPAC Name 5-methylbenzene-1,3-dicarboxylic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 298°C
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylisophthalic Acid
EINECSNumber (3)-1373
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.