Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylisoxazole, MP Biomedicals

Catalog No. ICN224768 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224768 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224768 Supplier MP Biomedicals, Inc Supplier No. 0522476810

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5765-44-6
Molecular Formula C4H5NO
Molecular Weight (g/mol) 83.09
InChI Key AGQOIYCTCOEHGR-UHFFFAOYSA-N
Synonym 5-methylisoxazole, isoxazole, 5-methyl, unii-yc2p4ibf1e, yc2p4ibf1e, 5-methyl isoxazole, 5-methylisoxazol, 5-methyl-isoxazole, pubchem8634, acmc-1arbg
PubChem CID 79833
IUPAC Name 5-methyl-1,2-oxazole
SMILES CC1=CC=NO1

Specifications

Quantity 10 g
Formula Weight 83.06
Chemical Name or Material 5-Methyl Isoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.