Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylpyridine-3-boronic acid, 95%, Thermo Scientific™

Catalog No. AC454272500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454272500 250 mg
AC454270010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454272500 Supplier Thermo Scientific Chemicals Supplier No. 454272500

Chemical Identifiers

CAS 173999-18-3
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.95
InChI Key REONQWGHSQHTAC-UHFFFAOYSA-N
Synonym 5-methylpyridine-3-boronic acid, 5-methyl-3-pyridineboronic acid, 5-methylpyridin-3-yl boronic acid, 5-methylpyridin-3-yl-3-boronic acid, 5-methylpyridine-3-boronicacid, 5-methyl-3-pyridyl boronic acid, 5-methyl-3-pyridinyl boronic acid, pubchem17082, 3-borono-5-methylpyridine, acmc-209e7n
PubChem CID 2762708
IUPAC Name (5-methylpyridin-3-yl)boronic acid
SMILES B(C1=CC(=CN=C1)C)(O)O

Specifications

Melting Point 300.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 136.95
Percent Purity 95%
Chemical Name or Material 5-Methylpyridine-3-boronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.