Learn More
Medchemexpress LLC 5-methylquinolin-8-ol | 5541-67-3 | MFCD00864652 | 99.6% | 159.19 | C10H9NO | 100 MG
SDP
Catalog No. 5000300990
Encompass_Preferred
Tiliquinol (5-methylquinolin-8-ol; NSC 130828) is a non-absorbed anti-amoebic luminal agent used in research to study Entamoeba infections and amebic liver abscess. It is supplied as a solid for laboratory use and is characterized by formula C10H9NO and molecular weight 159.19 g·mol-1.
  • Non-absorbed anti-amoebic luminal agent used for amebic research.
  • Applicable to studies of amebic liver abscess and related assays.
  • Chemical formula C10H9NO; molecular weight 159.19 g·mol-1.
  • Off-white to light yellow solid with reported purity of 99.55%.
  • Powder stability: -20°C for 3 years, 4°C for 2 years; in solvent: -80°C for 6 months.
  • Supplied in small solid quantities suitable for laboratory workflows.
Catalog No. 50-003-00990 Supplier Medchemexpress LLC Supplier No. HYW018397100MG
Add to Cart
Add to Cart
Tiliquinol (5-methylquinolin-8-ol; NSC 130828) is a non-absorbed anti-amoebic luminal agent used in research to study Entamoeba infections and amebic liver abscess. It is supplied as a solid for laboratory use and is characterized by formula C10H9NO and molecular weight 159.19 g·mol-1.
  • Non-absorbed anti-amoebic luminal agent used for amebic research.
  • Applicable to studies of amebic liver abscess and related assays.
  • Chemical formula C10H9NO; molecular weight 159.19 g·mol-1.
  • Off-white to light yellow solid with reported purity of 99.55%.
  • Powder stability: -20°C for 3 years, 4°C for 2 years; in solvent: -80°C for 6 months.
  • Supplied in small solid quantities suitable for laboratory workflows.