Learn More
Medchemexpress LLC 5-methylresorcinol monohydrate | 6153-39-5 | MFCD00149092 | 95.0% | 142.15 | C7H10O3 | 10 G
SDP
Catalog No. 5000300074
Encompass_Preferred
5-Methylresorcinol monohydrate (CAS 6153-39-5) is a research-grade crystalline solid with molecular formula C7H10O3 and molecular weight 142.15. The compound is supplied as a monohydrate with a reported purity of 95.0% and is used as a reagent or analytical standard in biochemical and enzymatic studies, including tyrosinase inhibition assays.
  • High purity suitable for research and analytical work.
  • Monohydrate crystalline solid for stable handling and storage.
  • Available in laboratory-scale package sizes for small-batch use.
  • Product specifications documented with datasheet, COA, and SDS.
  • Suitable as a reagent or reference standard in biochemical assays.
Catalog No. 50-003-00074 Supplier Medchemexpress LLC Supplier No. HYW01571510G
5-Methylresorcinol monohydrate (CAS 6153-39-5) is a research-grade crystalline solid with molecular formula C7H10O3 and molecular weight 142.15. The compound is supplied as a monohydrate with a reported purity of 95.0% and is used as a reagent or analytical standard in biochemical and enzymatic studies, including tyrosinase inhibition assays.
  • High purity suitable for research and analytical work.
  • Monohydrate crystalline solid for stable handling and storage.
  • Available in laboratory-scale package sizes for small-batch use.
  • Product specifications documented with datasheet, COA, and SDS.
  • Suitable as a reagent or reference standard in biochemical assays.