Learn More
Medchemexpress LLC 5-methylresorcinol monohydrate | 6153-39-5 | MFCD00149092 | 95.0% | 142.15 g/mol | C7H8O2·H2O | 5 G
SDP
Catalog No. 5000300014
Encompass_Preferred
5-Methylresorcinol monohydrate is a research-grade organic reagent supplied as a solid monohydrate and used in biochemical and pharmacological studies. It functions as a tyrosinase inhibitor and is used as a reference compound in enzyme assays and analytical methods.
  • High-purity solid suitable for research applications.
  • Acts as a tyrosinase inhibitor with reported cytotoxic and anxiolytic activity.
  • Monohydrate form with formula C7H8O2·H2O and molecular weight 142.15 g/mol.
  • Commonly used as a reference standard in enzyme assays and analytical methods.
  • Available in small laboratory pack sizes for bench research.
Catalog No. 50-003-00014 Supplier Medchemexpress LLC Supplier No. HYW0157155G
5-Methylresorcinol monohydrate is a research-grade organic reagent supplied as a solid monohydrate and used in biochemical and pharmacological studies. It functions as a tyrosinase inhibitor and is used as a reference compound in enzyme assays and analytical methods.
  • High-purity solid suitable for research applications.
  • Acts as a tyrosinase inhibitor with reported cytotoxic and anxiolytic activity.
  • Monohydrate form with formula C7H8O2·H2O and molecular weight 142.15 g/mol.
  • Commonly used as a reference standard in enzyme assays and analytical methods.
  • Available in small laboratory pack sizes for bench research.