Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-methylresorcinol monohydrate | 6153-39-5 | MFCD00149092 | 99.7% | 142.15 g/mol | C7H10O3 | 50 G
SDP

Catalog No. 5000301533
Encompass_Preferred

5-Methylresorcinol monohydrate (CAS 6153-39-5) is the monohydrate form of 5-methylresorcinol, supplied as a high-purity research reagent for biochemical and analytical applications. It is reported as a tyrosinase inhibitor and is used in laboratory studies of enzyme activity and small-molecule screening.

  • High purity (≈99.7%).
  • Monohydrate form for consistent stoichiometry.
  • Molecular weight 142.15 g/mol.
  • Chemical formula C7H10O3.
  • Reported tyrosinase inhibitor used in biochemical studies.
  • Suitable for analytical applications and assay development.
  • Available in laboratory pack sizes for research use.

Catalog No. 50-003-01533 Supplier Medchemexpress LLC Supplier No. HYW01571550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Methylresorcinol monohydrate (CAS 6153-39-5) is the monohydrate form of 5-methylresorcinol, supplied as a high-purity research reagent for biochemical and analytical applications. It is reported as a tyrosinase inhibitor and is used in laboratory studies of enzyme activity and small-molecule screening.

  • High purity (≈99.7%).
  • Monohydrate form for consistent stoichiometry.
  • Molecular weight 142.15 g/mol.
  • Chemical formula C7H10O3.
  • Reported tyrosinase inhibitor used in biochemical studies.
  • Suitable for analytical applications and assay development.
  • Available in laboratory pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.