Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylsalicylic Acid 98.0+%, TCI America™
SDP

Catalog No. C041025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C041025G 25 g
C04105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C041025G Supplier TCI America Supplier No. C041025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89-56-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002461
InChI Key DLGBEGBHXSAQOC-UHFFFAOYSA-N
Synonym 5-methylsalicylic acid, p-cresotic acid, p-cresotinic acid, 6-hydroxy-m-toluic acid, p-homosalicylic acid, 2,5-cresotic acid, benzoic acid, 2-hydroxy-5-methyl, 5-methyl-2-hydroxybenzoic acid, 6-hydroxy-3-methylbenzoic acid, alpha-cresotinic acid
PubChem CID 6973
IUPAC Name 2-hydroxy-5-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylsalicylic Acid
RTECSNumber GP3920200
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.