Learn More
Medchemexpress LLC HY-P7127 10ug Medchemexpress, Adiponectin (HEK293-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501970569
Encompass_Preferred
Medchemexpress, HY-113046 100mg 5-Methyltetrahydrofolic acid CAS:134-35-0 5-Methyltetrahydrofolic acid (5-Methyl THF) is a biologically active form of folic acid. 5-Methyltetrahydrofolic acid is a methylated derivate of tetrahydrofolate. 5-Methyltetrahydrofolic acid is the predominant natural dietary folate and the principal form of folate in plasma and cerebrospinal fluid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0569 Supplier Medchemexpress LLC Supplier No. HYP712710UG
Medchemexpress, HY-113046 100mg 5-Methyltetrahydrofolic acid CAS:134-35-0 5-Methyltetrahydrofolic acid (5-Methyl THF) is a biologically active form of folic acid. 5-Methyltetrahydrofolic acid is a methylated derivate of tetrahydrofolate. 5-Methyltetrahydrofolic acid is the predominant natural dietary folate and the principal form of folate in plasma and cerebrospinal fluid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.