Learn More
TARGETMOL CHEMICALS INC 5-METHYLTETRAHYDROFOLIC 500MG
SDP
Catalog No. 502577393
Encompass_Preferred
Also available in 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5-Methyltetrahydrofolic acid (5-Methyl THF) is a biologically active form of folic acid and a methylated derivate of tetrahydrofolate. It is the predominant natural dietary folate and the principal form of folate in plasma and cerebrospinal fluid. purity: 98%
Catalog No. 50-257-7393 Supplier TARGETMOL CHEMICALS INC Supplier No. T13516500MG
Also available in 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5-Methyltetrahydrofolic acid (5-Methyl THF) is a biologically active form of folic acid and a methylated derivate of tetrahydrofolate. It is the predominant natural dietary folate and the principal form of folate in plasma and cerebrospinal fluid. purity: 98%