Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylthio-1,3,4-thiadiazole-2-thiol, 97%, Thermo Scientific™

Catalog No. AAL0800206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0800206 5 g
AAL0800203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0800206 Supplier Thermo Scientific Chemicals Supplier No. L0800206

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6264-40-0
Molecular Formula C3H4N2S3
Molecular Weight (g/mol) 164.259
MDL Number MFCD00051648
InChI Key CFWGYKRJMYXYND-UHFFFAOYSA-N
Synonym 5-methylthio-1,3,4-thiadiazole-2-thiol, 2-mercapto-5-methylthio-1,3,4-thiadiazole, 1,3,4-thiadiazole-2 3h-thione, 5-methylthio, 5-methylsulfanyl-1,3,4-thiadiazole-2-thiol, 5-methylthio-1,3,4-thiadiazole-2 3h-thione, 5-methylsulfanyl-3h-1,3,4-thiadiazole-2-thione, maybridge1_000055, acmc-1b665, 2-methylthio-1,3,4-thiadiazole-5-thiol
PubChem CID 1241233
IUPAC Name 5-methylsulfanyl-3H-1,3,4-thiadiazole-2-thione
SMILES CSC1=NNC(=S)S1

Specifications

Melting Point 134°C to 139°C
Quantity 5 g
Beilstein 4009
Formula Weight 164.27
Percent Purity 97%
Chemical Name or Material 5-Methylthio-1,3,4-thiadiazole-2-thiol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 228-423-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.