Learn More
Cayman Chemical 5-Methyluridine, Purity: ≥98%, CAS: 1463-10-1, 5 g
SDP
Catalog No. 502939736
Encompass_Preferred
A pyrimidine nucleoside; enhances the antitumor activity of 5-fluorouracil in a mouse Erlich solid carcinoma model and a P388 murine leukemia model; has been used to characterize the activity of a variety of enzymes
Catalog No. 50-293-9736 Supplier Cayman Chemical Supplier No. 279865 G
A pyrimidine nucleoside; enhances the antitumor activity of 5-fluorouracil in a mouse Erlich solid carcinoma model and a P388 murine leukemia model; has been used to characterize the activity of a variety of enzymes