Learn More
Medchemexpress LLC 5-Methyluridine | 1463-10-1 | 99.9% | 258.23 | 500 MG
SDP
Catalog No. 5000366309
Encompass_Preferred
5-Methyluridine (m5U) is an RNA modified nucleotide generated by RNA methyltransferases (such as TrmA and RumA). It primarily targets specific uracil sites in RNA molecules like the T arm of tRNA and rRNA. 5-Methyluridine aids in physiological processes such as translation accuracy and ribosome function by stabilizing RNA folding or regulating base pairing through enzyme recognition of RNA secondary/tertiary structures.
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite
  • For research use only; not sold to patients
Catalog No. 50-003-66309 Supplier Medchemexpress LLC Supplier No. HYW009444500MG
5-Methyluridine (m5U) is an RNA modified nucleotide generated by RNA methyltransferases (such as TrmA and RumA). It primarily targets specific uracil sites in RNA molecules like the T arm of tRNA and rRNA. 5-Methyluridine aids in physiological processes such as translation accuracy and ribosome function by stabilizing RNA folding or regulating base pairing through enzyme recognition of RNA secondary/tertiary structures.
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite
  • For research use only; not sold to patients