Learn More
Medchemexpress LLC 5-Methyluridine | 1463-10-1 | MFCD00006535 | 99.88% | 258.23 | 1 ML
SDP
Catalog No. 5000453303
Encompass_Preferred
5-Methyluridine (m5U) is an RNA modified nucleotide generated by RNA methyltransferases (such as TrmA and RumA). It primarily targets specific uracil sites in RNA molecules like the T arm of tRNA and rRNA. 5-Methyluridine relies on enzyme recognition of RNA secondary/tertiary structures (such as the T loop of tRNA or the specific stem-loop structure of rRNA) and participates in physiological processes such as translation accuracy and ribosome function by stabilizing RNA folding or regulating base pairing.
  • Generated by RNA methyltransferases (such as TrmA and RumA).
  • Primarily targets specific uracil sites in RNA molecules like the T arm of tRNA and rRNA.
  • Participates in physiological processes such as translation accuracy and ribosome function.
  • Stabilizes RNA folding or regulates base pairing.
Catalog No. 50-004-53303 Supplier Medchemexpress LLC Supplier No. HYW00944410MM/1ML
5-Methyluridine (m5U) is an RNA modified nucleotide generated by RNA methyltransferases (such as TrmA and RumA). It primarily targets specific uracil sites in RNA molecules like the T arm of tRNA and rRNA. 5-Methyluridine relies on enzyme recognition of RNA secondary/tertiary structures (such as the T loop of tRNA or the specific stem-loop structure of rRNA) and participates in physiological processes such as translation accuracy and ribosome function by stabilizing RNA folding or regulating base pairing.
  • Generated by RNA methyltransferases (such as TrmA and RumA).
  • Primarily targets specific uracil sites in RNA molecules like the T arm of tRNA and rRNA.
  • Participates in physiological processes such as translation accuracy and ribosome function.
  • Stabilizes RNA folding or regulates base pairing.