Learn More
AnaSpec Beta - Amyloid (1 - 42), Human, 5mg
Supplier:  AnaSpec AS202765
Encompass
Beta-Amyloid (1-42), Human[DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA] ; MW 4514.1; (5 mg) AB (1-42), a major component of amyloid plaques, accumulates in neurons of Alzheimers disease brains. Biochemical analysis of the amyloid peptides isolated from Alzheimers disease brain indicates that AB (1-42) is the principal species associated with senile plaque amyloids, while AB (1-40) is more abundant in cerebrovascular amyloid deposit.
Catalog No. NC0833349