Learn More
Enzo Life Sciences Digoxigenin NHS ester, (5mg), CAS Number: 129273-26-3
SDP
Catalog No. NC1506157
Encompass_Preferred
Digoxigenin is a non-radioactive immune-tag, widely used for in situ hybridization and protein labeling. Pairing with anti-digoxigenin antibodies, digoxigenin tag allows the detection of interested biomolecules with high affinity and specificity. Digoxigenin NHS ester reacts with amino groups within biomolecules (nucleic acid, proteins, etc) and enables the attachment of digoxigenin into these molecules. Formula: C35H50N2O10. MW: 658.78. CAS Number: 129273-26-3. Purity: ≥80% (HPLC). Appearance: Off-white solid. Solubility: Soluble in DMSO or DMF. Use/Stability: Store, as supplied, at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C
Catalog No. NC1506157 Supplier Enzo Life Sciences Supplier No. ENZ450220005
Digoxigenin is a non-radioactive immune-tag, widely used for in situ hybridization and protein labeling. Pairing with anti-digoxigenin antibodies, digoxigenin tag allows the detection of interested biomolecules with high affinity and specificity. Digoxigenin NHS ester reacts with amino groups within biomolecules (nucleic acid, proteins, etc) and enables the attachment of digoxigenin into these molecules. Formula: C35H50N2O10. MW: 658.78. CAS Number: 129273-26-3. Purity: ≥80% (HPLC). Appearance: Off-white solid. Solubility: Soluble in DMSO or DMF. Use/Stability: Store, as supplied, at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C