Learn More
Bio X Cell Anti-Human CD8a, Clone OKT-8, IgG2a, 5 MG
Supplier: Bio X Cell BE00042R005MG
The OKT-8 monoclonal antibody reacts with human CD8α. CD8 is a transmembrane glycoprotein that functions as a co-receptor for the T cell receptor (TCR) and is primarily expressed on cytotoxic T cells, thymocytes, natural killer cells, and some dendritic cell subsets.
- Clone: OKT-8
- Reactivities: Human
- Isotype: Mouse IgG2a
- Immunogen: Human CD8α
- Reported applications: In vivo CD8+ T cell depletion in humanized mice
- Formulation: PBS, pH 6.0, 0.01% Tween
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.