Learn More
Labcon North America SuperClear 0.5mL Freestanding Screw Cap Microcentrifuge Tubes with Caps, Sterile
Catalog No. NC0867920
Encompass_Preferred
Tube, Microcentrifuge; Screw Cap Style; Labcon; SuperClear; 0.5mL; Freestanding with attached flat screw caps; Caps include integral ethylene O-ring; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 20,000 x G; Freezable -80C; Autoclavable; Capacity: 0.5mL; Natural color caps; 500 tubes per bag/10 bags per case; Made in USA
Catalog No. NC0867920 Supplier Labcon North America Supplier No. 36418750009
Tube, Microcentrifuge; Screw Cap Style; Labcon; SuperClear; 0.5mL; Freestanding with attached flat screw caps; Caps include integral ethylene O-ring; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 20,000 x G; Freezable -80C; Autoclavable; Capacity: 0.5mL; Natural color caps; 500 tubes per bag/10 bags per case; Made in USA