Learn More
APPLIED INDUSTRIAL TECHNOLOGY 5-Minute Epoxy Gel, 25 mL
Catalog No. NC9572975
Encompass
Thixotropic, nonmigrating gel adhesive with excellent gap-filling properties. Cures in one hour.
Catalog No. NC9572975 Supplier APPLIED INDUSTRIAL TECHNOLOGY Supplier No. 1424025ML
Thixotropic, nonmigrating gel adhesive with excellent gap-filling properties. Cures in one hour.