Learn More
Takara Bio 5 ML MAG-TRYPSIN
SDP
Catalog No. NC9618662
Encompass
MS-grade trypsin (TPCK-trypsin) immobilized on magnetic beads for effective trypsin digestion. TPCK-tryspin shows no chymotryptic and minimal autolytic activity. No centrifugation required.
Catalog No. NC9618662 Supplier Takara Bio Supplier No. 635646
MS-grade trypsin (TPCK-trypsin) immobilized on magnetic beads for effective trypsin digestion. TPCK-tryspin shows no chymotryptic and minimal autolytic activity. No centrifugation required.