Learn More
USA Scientific Inc 5 mL serological pipet
Catalog No. 502745296
Encompass
5 mL serological pipet, 0.1 mL graduations, standard tip, sterile, 25/sleeve, 500/case
Catalog No. 50-274-5296 Supplier USA Scientific Inc Supplier No. 56606107
5 mL serological pipet, 0.1 mL graduations, standard tip, sterile, 25/sleeve, 500/case