Learn More
Genesee Scientific Corporation Genesee Scientific 5 ML Serological Pipets, Sterile, DNase/RNase Free, Optical Grade Polystyrene, 5 ml
SDP
Catalog No. NC0631030
Encompass
The Genesee Scientific 5 ML Serological Pipets are designed for accurate liquid handling in cell and tissue culture. Made from optical grade polystyrene, these sterile pipets feature bidirectional graduations and a patented filter for enhanced barrier protection. Manufactured in a Class 100,000 cleanroom, they meet stringent quality standards including DNase/RNase Free certifications, ensuring reliability and precision in laboratory applications.
Catalog No. NC0631030 Supplier Genesee Scientific Corporation Supplier No. 12102
The Genesee Scientific 5 ML Serological Pipets are designed for accurate liquid handling in cell and tissue culture. Made from optical grade polystyrene, these sterile pipets feature bidirectional graduations and a patented filter for enhanced barrier protection. Manufactured in a Class 100,000 cleanroom, they meet stringent quality standards including DNase/RNase Free certifications, ensuring reliability and precision in laboratory applications.