Learn More
USA Scientific Inc 5 mL transfer pipet, grad
Catalog No. 502745394
Encompass
5 mL polyethylene transfer pipet, graduated to 1 mL, bulb draw 3.5 mL, length 15.0 cm. 22 Drops per mL, individually wrapped, 500/case
Catalog No. 50-274-5394 Supplier USA Scientific Inc Supplier No. 10201111
5 mL polyethylene transfer pipet, graduated to 1 mL, bulb draw 3.5 mL, length 15.0 cm. 22 Drops per mL, individually wrapped, 500/case