Learn More
NEST SCIENTIFIC INC 5 ML VIAL 2.5 ML VTM STE 100/C
Catalog No. NC2887674
Encompass_Preferred
FDA, 510k (K211256) approved Viral Transport Media Kit. Includes a leakrpoof 5mL transport tube rated to 95kPa filled with 2.5mL of VTM. Includes flocked nylon nasopharyngeal swabs and sealing biohazard transport bags. VTM stabilizes viral specimens at 4C for 48 hours. Unused VTM can be stored at room temperature. Produced in a cGMP (21 CFR part 820/821), ISO 13485 and ISO 9001 facility.
Catalog No. NC2887674 Supplier NEST SCIENTIFIC INC Supplier No. 202115
FDA, 510k (K211256) approved Viral Transport Media Kit. Includes a leakrpoof 5mL transport tube rated to 95kPa filled with 2.5mL of VTM. Includes flocked nylon nasopharyngeal swabs and sealing biohazard transport bags. VTM stabilizes viral specimens at 4C for 48 hours. Unused VTM can be stored at room temperature. Produced in a cGMP (21 CFR part 820/821), ISO 13485 and ISO 9001 facility.