Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Sodium Chloride (5M), 10 Liters
SDP

Catalog No. 50146830
Encompass_Preferred

Sodium Chloride (5M) is suitable for molecular biology, protein chemistry and biochemical applications.

5M concentration
Filtered through a 0.22 µm membrane
DNase, RNase, and Protease tested

Applications: Sodium Chloride (5M) is free of detectable endonucleases, ribonucleases and nicking activity according to the current quality control procedures.

Storage Condition: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase, and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-146-830 Supplier Quality Biological Inc Supplier No. 351036151
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Chloride (5M) is suitable for molecular biology, protein chemistry and biochemical applications.

5M concentration
Filtered through a 0.22 µm membrane
DNase, RNase, and Protease tested

Applications: Sodium Chloride (5M) is free of detectable endonucleases, ribonucleases and nicking activity according to the current quality control procedures.

Storage Condition: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase, and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.