Learn More
Medchemexpress LLC 5-(morpholine-4-carbonyl)-2,1,3-benzoxadiazole | 867276-98-0 | 99.9% | 233.22 g·mol⁻1 | C11H11N3O3 | 5 MG
SDP
Catalog No. 5000221670
Encompass_Preferred
CX 717 is a small-molecule positive allosteric modulator of AMPA receptors used in preclinical research for cognitive enhancement, antidepressant-like effects, and attention-deficit/hyperactivity disorder (ADHD) studies. It is supplied as a white to off-white solid with high purity for laboratory use and requires storage protected from light at refrigerated temperatures.
  • Positive allosteric modulator of AMPA receptors.
  • Shows antidepressant-like activity in preclinical models.
  • Used in research on cognition and ADHD-related mechanisms.
  • High purity suitable for analytical and biological assays.
  • Solid form, white to off-white, stable under recommended storage.
Catalog No. 50-002-21670 Supplier Medchemexpress LLC Supplier No. HY1398975MG
CX 717 is a small-molecule positive allosteric modulator of AMPA receptors used in preclinical research for cognitive enhancement, antidepressant-like effects, and attention-deficit/hyperactivity disorder (ADHD) studies. It is supplied as a white to off-white solid with high purity for laboratory use and requires storage protected from light at refrigerated temperatures.
  • Positive allosteric modulator of AMPA receptors.
  • Shows antidepressant-like activity in preclinical models.
  • Used in research on cognition and ADHD-related mechanisms.
  • High purity suitable for analytical and biological assays.
  • Solid form, white to off-white, stable under recommended storage.