Learn More
Medchemexpress LLC 5-(morpholine-4-carbonyl)-2,1,3-benzoxadiazole | 867276-98-0 | 99.9% | C11H11N3O3 | 10MG
SDP
Catalog No. 5000261276
Encompass_Preferred
CX 717 is a small-molecule positive allosteric modulator (PAM) of AMPA receptors used in preclinical research. It exhibits antidepressant-like effects and is applied in studies of cognition and adult attention deficit hyperactivity disorder (ADHD). The compound is provided as a high-purity research chemical available in solid form and as a DMSO solution.
  • Positive allosteric modulator of AMPA receptors.
  • Demonstrates antidepressant-like effects in preclinical studies.
  • Used in research of cognition and adult ADHD.
  • High purity (99.9%).
  • Available as solid and as DMSO solution.
  • Recommended storage: 4°C; in solvent store at -80°C for long term.
Catalog No. 50-002-61276 Supplier Medchemexpress LLC Supplier No. HY13989710MG
CX 717 is a small-molecule positive allosteric modulator (PAM) of AMPA receptors used in preclinical research. It exhibits antidepressant-like effects and is applied in studies of cognition and adult attention deficit hyperactivity disorder (ADHD). The compound is provided as a high-purity research chemical available in solid form and as a DMSO solution.
  • Positive allosteric modulator of AMPA receptors.
  • Demonstrates antidepressant-like effects in preclinical studies.
  • Used in research of cognition and adult ADHD.
  • High purity (99.9%).
  • Available as solid and as DMSO solution.
  • Recommended storage: 4°C; in solvent store at -80°C for long term.