Learn More
Medchemexpress LLC 5-(N,N-Hexamethylene)-amiloride | 1428-95-1 | 99.1% | 1 ML
SDP
Catalog No. 5000339229
Encompass_Preferred
5-(N,N-Hexamethylene)-amiloride, also known as Hexamethylene amiloride (HMA), is a potent Na+/H+ exchanger inhibitor derived from amiloride. It effectively decreases intracellular pH (pHi) and has been shown to induce apoptosis in leukemic cells. This compound also acts as an inhibitor of the HIV-1 Vpu virus ion channel and inhibits the replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E) in cultured L929 cells.
  • Potent Na+/H+ exchanger inhibitor
  • Decreases intracellular pH (pHi)
  • Induces apoptosis in leukemic cells
  • Inhibits HIV-1 Vpu virus ion channel
  • Inhibits replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E)
  • Inhibits human cardiac ion channels hERG, Nav1.5, and Cav1.2
  • Exhibits microsomal and mouse plasma stability
  • Displays Caco-2 cell permeability
Catalog No. 50-003-39229 Supplier Medchemexpress LLC Supplier No. HY12806710MM/1ML
5-(N,N-Hexamethylene)-amiloride, also known as Hexamethylene amiloride (HMA), is a potent Na+/H+ exchanger inhibitor derived from amiloride. It effectively decreases intracellular pH (pHi) and has been shown to induce apoptosis in leukemic cells. This compound also acts as an inhibitor of the HIV-1 Vpu virus ion channel and inhibits the replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E) in cultured L929 cells.
  • Potent Na+/H+ exchanger inhibitor
  • Decreases intracellular pH (pHi)
  • Induces apoptosis in leukemic cells
  • Inhibits HIV-1 Vpu virus ion channel
  • Inhibits replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E)
  • Inhibits human cardiac ion channels hERG, Nav1.5, and Cav1.2
  • Exhibits microsomal and mouse plasma stability
  • Displays Caco-2 cell permeability