Learn More
Medchemexpress LLC 5-(N,N-hexamethylene)-amiloride | 1428-95-1 | 99.1% | 10 MG
SDP
Catalog No. 5000339496
Encompass_Preferred
5-(N,N-Hexamethylene)-amiloride, also known as HMA, is a potent amiloride derivative that functions as an inhibitor of the Na+/H+ exchanger. This action decreases intracellular pH and induces apoptosis in leukemic cells. It also acts as an inhibitor of the HIV-1 Vpu virus ion channel, demonstrating efficacy against the replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E) in cultured L929 cells. This product is intended for research use only.
  • Potent Na+/H+ exchanger inhibitor
  • Induces apoptosis in leukemic cells
  • Inhibits HIV-1 Vpu virus ion channel
  • Inhibits replication of mouse hepatitis virus (MHV)
  • Inhibits replication of human coronavirus 229E (HCoV229E)
  • High purity (99.1%)
Catalog No. 50-003-39496 Supplier Medchemexpress LLC Supplier No. HY12806710MG
5-(N,N-Hexamethylene)-amiloride, also known as HMA, is a potent amiloride derivative that functions as an inhibitor of the Na+/H+ exchanger. This action decreases intracellular pH and induces apoptosis in leukemic cells. It also acts as an inhibitor of the HIV-1 Vpu virus ion channel, demonstrating efficacy against the replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E) in cultured L929 cells. This product is intended for research use only.
  • Potent Na+/H+ exchanger inhibitor
  • Induces apoptosis in leukemic cells
  • Inhibits HIV-1 Vpu virus ion channel
  • Inhibits replication of mouse hepatitis virus (MHV)
  • Inhibits replication of human coronavirus 229E (HCoV229E)
  • High purity (99.1%)