Learn More
Medchemexpress LLC 5-(N,N-Hexamethylene)-amiloride | 1428-95-1 | >99.1% | 100 MG
SDP
Catalog No. 5000342337
Encompass_Preferred
5-(N,N-Hexamethylene)-amiloride (HMA) is a potent Na+/H+ exchanger inhibitor derived from amiloride. It effectively decreases intracellular pH and induces apoptosis in leukemic cells. Additionally, it functions as an inhibitor of the HIV-1 Vpu virus ion channel and demonstrates inhibitory effects on the replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E) in cultured L929 cells.
  • Potent Na+/H+ exchanger inhibitor
  • Decreases intracellular pH
  • Induces apoptosis in leukemic cells
  • Inhibits HIV-1 Vpu virus ion channel
  • Inhibits mouse hepatitis virus (MHV) replication
  • Inhibits human coronavirus 229E (HCoV229E) replication
  • Purity of >99.1%
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-003-42337 Supplier Medchemexpress LLC Supplier No. HY128067100MG
5-(N,N-Hexamethylene)-amiloride (HMA) is a potent Na+/H+ exchanger inhibitor derived from amiloride. It effectively decreases intracellular pH and induces apoptosis in leukemic cells. Additionally, it functions as an inhibitor of the HIV-1 Vpu virus ion channel and demonstrates inhibitory effects on the replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E) in cultured L929 cells.
  • Potent Na+/H+ exchanger inhibitor
  • Decreases intracellular pH
  • Induces apoptosis in leukemic cells
  • Inhibits HIV-1 Vpu virus ion channel
  • Inhibits mouse hepatitis virus (MHV) replication
  • Inhibits human coronavirus 229E (HCoV229E) replication
  • Purity of >99.1%
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year