Learn More
Medchemexpress LLC 5-(N,N-Hexamethylene)-amiloride | 1428-95-1 | 99.14% | 311.77 | 5 MG
SDP
Catalog No. 5000364391
Encompass_Preferred
5-(N,N-Hexamethylene)-amiloride (Hexamethylene amiloride) is a derivative of amiloride, functioning as a potent Na+/H+ exchanger inhibitor. It effectively decreases intracellular pH and induces apoptosis in leukemic cells. Furthermore, it inhibits the HIV-1 Vpu virus ion channel and has been shown to suppress the replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E) in cultured L929 cells.
  • Potent Na+/H+ exchanger inhibitor
  • Decreases intracellular pH
  • Induces apoptosis in leukemic cells
  • Inhibits HIV-1 Vpu virus ion channel
  • Suppresses mouse hepatitis virus (MHV) replication
  • Suppresses human coronavirus 229E (HCoV229E) replication
  • Inhibits human cardiac ion channels hERG, Nav1.5, and Cav1.2
  • Exhibits microsomal stability
  • Shows mouse plasma stability and plasma protein binding
  • Displays Caco-2 cell permeability
Catalog No. 50-003-64391 Supplier Medchemexpress LLC Supplier No. 5MGHY128067
5-(N,N-Hexamethylene)-amiloride (Hexamethylene amiloride) is a derivative of amiloride, functioning as a potent Na+/H+ exchanger inhibitor. It effectively decreases intracellular pH and induces apoptosis in leukemic cells. Furthermore, it inhibits the HIV-1 Vpu virus ion channel and has been shown to suppress the replication of mouse hepatitis virus (MHV) and human coronavirus 229E (HCoV229E) in cultured L929 cells.
  • Potent Na+/H+ exchanger inhibitor
  • Decreases intracellular pH
  • Induces apoptosis in leukemic cells
  • Inhibits HIV-1 Vpu virus ion channel
  • Suppresses mouse hepatitis virus (MHV) replication
  • Suppresses human coronavirus 229E (HCoV229E) replication
  • Inhibits human cardiac ion channels hERG, Nav1.5, and Cav1.2
  • Exhibits microsomal stability
  • Shows mouse plasma stability and plasma protein binding
  • Displays Caco-2 cell permeability