Learn More
Cayman Chemical 5-(N,N-hexamethylene)-Amiloride, Purity: ≥98%, CAS: 1428-95-1, 50 mg
SDP
Catalog No. 502990219
Encompass_Preferred
An amiloride derivative with diverse biological activities; an allosteric antagonist of adenosine A2A receptors (Ki = 3.3 µM); inhibits the cation-selective ion channel formed by Vpu at 50 µM, as well as budding of virus-like particles in HeLa cells expressing the HIV-1 proteins Gag and Vpu at 10 µM; blocks the cation-selective ion channels formed by p7; induces necrosis in and reduces the viability of various breast cancer cells at 40 µM; protects against post-ischemic contractile dysfunction and reduces coronary effluent creatine phosphokinase activity in a model of ischemia-reperfusion injury using isolated rat right ventricular free walls
Catalog No. 50-299-0219 Supplier Cayman Chemical Supplier No. 2978850 MG
An amiloride derivative with diverse biological activities; an allosteric antagonist of adenosine A2A receptors (Ki = 3.3 µM); inhibits the cation-selective ion channel formed by Vpu at 50 µM, as well as budding of virus-like particles in HeLa cells expressing the HIV-1 proteins Gag and Vpu at 10 µM; blocks the cation-selective ion channels formed by p7; induces necrosis in and reduces the viability of various breast cancer cells at 40 µM; protects against post-ischemic contractile dysfunction and reduces coronary effluent creatine phosphokinase activity in a model of ischemia-reperfusion injury using isolated rat right ventricular free walls