Learn More
Cayman Chemical 5 NCyclopropylcarboxamIdo 10mg
SDP
Catalog No. 502966545
Encompass_Preferred
A specific adenosine A2 receptor agonist with antipyretic and anticonvulsant activity; stimulates the production of cyclic AMP in CHO-K1 cells with an EC50 value of 5.3 µM
Catalog No. 50-296-6545 Supplier Cayman Chemical Supplier No. 1815710 MG
A specific adenosine A2 receptor agonist with antipyretic and anticonvulsant activity; stimulates the production of cyclic AMP in CHO-K1 cells with an EC50 value of 5.3 µM