Learn More
Cayman Chemical 5'-(N-Cyclopropyl)carboxamidoadenosine, Purity: ≥95%, CAS: 50908-62-8, 50 mg
SDP
Catalog No. 502966678
Encompass_Preferred
A specific adenosine A2 receptor agonist with antipyretic and anticonvulsant activity; stimulates the production of cyclic AMP in CHO-K1 cells with an EC50 value of 5.3 µM
Catalog No. 50-296-6678 Supplier Cayman Chemical Supplier No. 1815750 MG
A specific adenosine A2 receptor agonist with antipyretic and anticonvulsant activity; stimulates the production of cyclic AMP in CHO-K1 cells with an EC50 value of 5.3 µM