Learn More
Cayman Chemical 5-ndrostentrIol 10mg
SDP
Catalog No. 502910000
Encompass_Preferred
An active metabolite of DHEA; reduces radiation-induced lethality and induces hematopoietic recovery from surviving stem cells in mice at 0.75 mg/25 g body weight; increases survival in a rat model of surgical trauma-induced sepsis at 40 mg/kg; urinary levels are increased in women with androgenic alopecia
Catalog No. 50-291-0000 Supplier Cayman Chemical Supplier No. 3527310 MG
An active metabolite of DHEA; reduces radiation-induced lethality and induces hematopoietic recovery from surviving stem cells in mice at 0.75 mg/25 g body weight; increases survival in a rat model of surgical trauma-induced sepsis at 40 mg/kg; urinary levels are increased in women with androgenic alopecia