Learn More
Cayman Chemical 5 NethylNIsopropylAmIlo 10mg
SDP
Catalog No. 502971423
Encompass_Preferred
A potent inhibitor of several NHE isoforms, inhibiting NHE1, NHE2, NHE3, and NHE5 with Ki values of 0.02, 0.5, 2.4, and 0.42 μM, respectively; less effectively inhibits NHE4 (IC50 ≥10 μM)
Catalog No. 50-297-1423 Supplier Cayman Chemical Supplier No. 1440610 MG
A potent inhibitor of several NHE isoforms, inhibiting NHE1, NHE2, NHE3, and NHE5 with Ki values of 0.02, 0.5, 2.4, and 0.42 μM, respectively; less effectively inhibits NHE4 (IC50 ≥10 μM)