Learn More
5-Nitro-2-furoic acid, 98+%
Description
5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 186°C to 188°C |
| Quantity | 5 g |
| Beilstein | 139373 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 157.08 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 5-Nitro-2-furoic acid |
Safety and Handling
| Hazard Category | H315-H319-H335-H341 |
| Hazard Statement | GHS H Statement H341 Suspected of causing genetic defects. |
| Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c |
| EINECSNumber | 211-432-6 |
| RTECSNumber | LV2012000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.