Learn More
Medchemexpress LLC 5-nitrobenzimidazole | 94-52-0 | 99.7% | 163.14 | 25 G
SDP
Catalog No. 5000452407
Encompass_Preferred
5-nitrobenzimidazole is a genotoxic nitro compound and a 6-aminopurine analogue. It exhibits a slight inhibitory effect on xanthine oxidase, making it suitable for research related to hyperuricemia.
  • Genotoxic nitro compound
  • 6-aminopurine analogue
  • Slight inhibitory effect on xanthine oxidase
  • Suitable for hyperuricemia research
Catalog No. 50-004-52407 Supplier Medchemexpress LLC Supplier No. 25GHYW010132
5-nitrobenzimidazole is a genotoxic nitro compound and a 6-aminopurine analogue. It exhibits a slight inhibitory effect on xanthine oxidase, making it suitable for research related to hyperuricemia.
  • Genotoxic nitro compound
  • 6-aminopurine analogue
  • Slight inhibitory effect on xanthine oxidase
  • Suitable for hyperuricemia research