Learn More
Medchemexpress LLC 5-Nitrobenzimidazole | 94-52-0 | 99.7% | 163.14 | 50 G
SDP
Catalog No. 5000453906
Encompass_Preferred
5-nitrobenzimidazole is a genotoxic nitro compound and a 6-aminopurine analogue. It demonstrates a slight inhibitory effect on xanthine oxidase and can be utilized in hyperuricemia research.
  • Used in hyperuricemia research
  • Shows a slight inhibitory effect on xanthine oxidase
Catalog No. 50-004-53906 Supplier Medchemexpress LLC Supplier No. HYW01013250G
5-nitrobenzimidazole is a genotoxic nitro compound and a 6-aminopurine analogue. It demonstrates a slight inhibitory effect on xanthine oxidase and can be utilized in hyperuricemia research.
  • Used in hyperuricemia research
  • Shows a slight inhibitory effect on xanthine oxidase