Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitrofluorescein (isomer I) 98.0+%, TCI America™
SDP

Catalog No. N01991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01991G 1 g
N01995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N01991G Supplier TCI America Supplier No. N01991G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3326-35-0
Molecular Formula C20H11NO7
MDL Number MFCD00135108

Specifications

Quantity 1 g
Synonym 5-Nitro-3′C,6′C-dihydroxyspiro[isobenzofuran-1(3H),9′C-(9H)xanthen]-3-one
Formula Weight 377.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitrofluorescein (isomer I)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.