Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroguaiacol 98.0+%, TCI America™
SDP

Catalog No. N0498500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0498500G 500 g
N049825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0498500G Supplier TCI America Supplier No. N0498500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 636-93-1
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.136
MDL Number MFCD00015561
InChI Key KXKCTSZYNCDFFG-UHFFFAOYSA-N
Synonym 5-nitroguaiacol, phenol, 2-methoxy-5-nitro, 3-hydroxy-4-methoxynitrobenzene, 2-methoxy-5-nitro-phenol, unii-e5t11i2jub, e5t11i2jub, 2-hydroxy-4-nitroanisole, acmc-1b3lw, 2-methoxy-5-nitro phenol, 2-methyloxy-5-nitrophenol
PubChem CID 69471
ChEBI CHEBI:81049
IUPAC Name 2-methoxy-5-nitrophenol
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])O

Specifications

Melting Point 105°C
Boiling Point 112°C
Quantity 500 g
Formula Weight 169.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroguaiacol
EINECSNumber (3)-2771&(9)-0970
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.