Learn More
Matrix Scientific 5-Nitroindole-2-carboxylic acid, 16730-20-4, MFCD00236372, 1g
Supplier: Matrix Scientific 0126031G
Molecular Formula C9H6N2O4, Purity 98%, Molecular Weight 206.16, Melting Point 326-328°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.