Learn More
Medchemexpress LLC 5-Nitroindole | 6146-52-7 | 98.6% | C8H6N2O2 | 10 G
SDP
Catalog No. 5000434216
Encompass_Preferred
5-Nitroindole is a mutagenic nitroarene and universal base analog. It significantly reduces the cellular adenylate energy charge by decreasing ATP levels and increasing AMP levels, and inhibits lipid peroxidation.
  • Decreases adenylate energy charge in rat hepatocytes
  • Inhibits NADPH-dependent, t-BuOOH-dependent, and NADH-initiated lipid peroxidation
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-34216 Supplier Medchemexpress LLC Supplier No. HY5927310G
5-Nitroindole is a mutagenic nitroarene and universal base analog. It significantly reduces the cellular adenylate energy charge by decreasing ATP levels and increasing AMP levels, and inhibits lipid peroxidation.
  • Decreases adenylate energy charge in rat hepatocytes
  • Inhibits NADPH-dependent, t-BuOOH-dependent, and NADH-initiated lipid peroxidation
  • Soluble in DMSO at 100 mg/mL