Learn More
Medchemexpress LLC 5-Nitroindole | 6146-52-7 | MFCD00005673 | >99.3% | C8H6N2O2 | 25 G
SDP
Catalog No. 5000359046
Encompass_Preferred
5-Nitroindole is a mutagenic nitroarene and a universal base analog. It functions by significantly reducing cellular adenylate energy charge through decreasing ATP levels and increasing AMP levels, and effectively inhibits lipid peroxidation in liver microsomes and mitochondria.
  • Reduces cellular adenylate energy charge
  • Inhibits lipid peroxidation
  • Available as a yellow to green solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for solutions: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-59046 Supplier Medchemexpress LLC Supplier No. 25GHY59273
5-Nitroindole is a mutagenic nitroarene and a universal base analog. It functions by significantly reducing cellular adenylate energy charge through decreasing ATP levels and increasing AMP levels, and effectively inhibits lipid peroxidation in liver microsomes and mitochondria.
  • Reduces cellular adenylate energy charge
  • Inhibits lipid peroxidation
  • Available as a yellow to green solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for solutions: -80°C for 6 months, -20°C for 1 month