Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroindole, 99%

Catalog No. p-7080875
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1060803 1 g
AAA1060806 5 g
AAA1060814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1060803 Supplier Thermo Scientific Chemicals Supplier No. A1060803
Only null left
Add to Cart

CAS: 6146-52-7 | C8H6N2O2 | 162.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6146-52-7
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00005673
InChI Key OZFPSOBLQZPIAV-UHFFFAOYSA-N
Synonym 5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr
PubChem CID 22523
IUPAC Name 5-nitro-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1[N+](=O)[O-]

Specifications

Melting Point 139°C to 143°C
Quantity 1 g
Beilstein 383777
Formula Weight 162.15
Percent Purity 99%
Chemical Name or Material 5-Nitroindole
Hazard Category H302-H319-H340
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
EINECSNumber 228-153-0
RTECSNumber NM1168200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.