Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroquinoline 98.0+%, TCI America™
SDP

Catalog No. N034425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N034425G 25 g
N0344250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N034425G Supplier TCI America Supplier No. N034425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 607-34-1
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00006790
InChI Key NDDZXHOCOKCNBM-UHFFFAOYSA-N
Synonym quinoline, 5-nitro, 5-nitro-quinoline, zlchem 399, pubchem5821, acmc-209mlr, 5-nitroquinoline, 5-20-07-00325 beilstein handbook reference, ksc355k8h, 7ze
PubChem CID 11829
IUPAC Name 5-nitroquinoline
SMILES C1=CC2=C(C=CC=N2)C(=C1)[N+](=O)[O-]

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 25 g
Formula Weight 174.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroquinoline
RTECSNumber VC1850000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.