Learn More
Cayman Chemical 5-Nitroso-8-quinolinol, Purity: ≥95%, CAS: 3565-26-2, 10 mg
SDP
Catalog No. 502955284
Encompass_Preferred
An HDAC inhibitor, antitumor agent that at 10 μM can induce oxidative stress contributing to apoptosis and differentiation in MCF-7 breast cancer cells; inhibits T. gondii tachyzoite propagation in human fibroblasts (EC50 = 80 nM) and inhibits P. falciparum growth in human red blood cells (EC50 = 1.3 μM)
Catalog No. 50-295-5284 Supplier Cayman Chemical Supplier No. 1385610 MG
An HDAC inhibitor, antitumor agent that at 10 μM can induce oxidative stress contributing to apoptosis and differentiation in MCF-7 breast cancer cells; inhibits T. gondii tachyzoite propagation in human fibroblasts (EC50 = 80 nM) and inhibits P. falciparum growth in human red blood cells (EC50 = 1.3 μM)